Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Deferoxamine mesylate | 138-14-7 | MFCD00058605 | 99.5% | 656.79 | C26H52N6O11S | 10 MG
SDP

Supplier:  Medchemexpress LLC HYB0988R10MG

Encompass_Preferred

Deferoxamine mesylate is an iron-chelating research standard used as a hypoxia mimetic and to chelate ferric ions in biochemical and cell biology studies. Provided as a solid for laboratory use; intended for research purposes only.

  • Iron chelator and hypoxia mimetic for biochemical and cell biology studies.
  • CAS number 138-14-7.
  • Molecular weight 656.79 g/mol.
  • Purity approximately 99.5% (per certificate of analysis).
  • Supplied as a solid powder suitable for preparing stock solutions.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-98213


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.