Learn More
Selleck Chemical LLC Deferoxamine mesylate
SDP
Supplier:  Selleck Chemical LLC S57421G
Encompass_Preferred
Deferoxamine mesylate is the mesylate salt of Deferoxamine which forms iron complexes and is used as a chelating agent Deferoxamine is a ferroptosis inhibitor that stabilizes HIF-1 expression and improves HIF-1 transactivity in hypoxic and hyperglycemic states in vitro Deferoxamine decreases beta-amyloid (A ) deposition and induces autophagy Please do not prepare stock solutions with normal saline or PBS as precipitation may occur
Catalog No. 50-000-03088