Learn More
Selleck Chemical LLC Deferoxamine mesylate 25mg 138-14-7 Desferrioxamine B
SDP
Supplier:  Selleck Chemical LLC S574225MG
Encompass_Preferred
Deferoxamine mesylate (Desferrioxamine B) is the mesylate salt of Deferoxamine, which forms iron complexes and is used as a chelating agent. Deferoxamine is a ferroptosis inhibitor that stabilizes HIF-1? expression and improves HIF-1? transactivity in hypoxic and hyperglycemic states in vitro. Deferoxamine decreases beta-amyloid (A?) deposition and induces autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8113