Learn More
Apexbio Technology LLC Deferoxamine mesylate 138-14-7 500mg
SDP
Supplier:  Apexbio Technology LLC B6068500
Encompass_Preferred
Deferoxamine mesylate (CAS 138-14-7) is an iron-specific chelating agent used primarily to manage acute iron poisoning It binds iron ions to form a water-soluble complex ferrioxamine thus preventing iron from participating in harmful oxidative reactions and facilitating renal excretion In rat mammary adenocarcinoma models deferoxamine mesylate treatment particularly combined with low-iron diet reduces tumor growth Additionally preoperative administration attenuates oxidative stress markers and pancreatic tissue damage by inducing HIF-1 expression in rat liver transplantation studies highlighting its utility in anticancer and cytoprotective research
Catalog No. 50-000-63588