Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Deflazacort-d3 | 2280940-17-0 | 444.54 | C25H28D3NO6 | 1 MG
SDP

Supplier:  Medchemexpress LLC HYW6543971MG

Encompass_Preferred

Deflazacort-d3 is a deuterated isotope-labeled analog of deflazacort used as an analytical internal standard for quantitative assays and mass spectrometry. Supplied in small milligram pack sizes for laboratory research; consult the certificate of analysis for purity and storage instructions.

  • Deuterated internal standard for accurate quantitation in LC-MS/MS assays.
  • Offered in milligram quantities suitable for analytical workflows.
  • Molecular formula C25H28D3NO6; molecular weight 444.54.
  • CAS number 2280940-17-0 for unambiguous identification.
  • Datasheet and handling instructions available; check CoA for purity.

Catalog No. 50-002-84493


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.