Learn More
Selleck Chemical LLC DEG-35-E1412-5MG
SDP
Supplier:  Selleck Chemical LLC E14125MG
Encompass_Preferred
DEG-35 is a cereblon-dependent degrader of IKZF2 and casein kinase 1 (CK1 ) It blocks cell growth and delays leukemia progression in murine and human acute myeloid leukemia (AML) mouse models
Catalog No. 50-313-4465