Learn More
Selleck Chemical LLC DEG-77-E1413-5MG
SDP
Supplier:  Selleck Chemical LLC E14135MG
Encompass_Preferred
DEG-77 is a cereblon-dependent degrader of IKZF2 and casein kinase 1 (CK1 ) It blocks cell growth and delays leukemia progression in murine and human acute myeloid leukemia (AML) mouse models
Catalog No. 50-313-4466