Learn More
Selleck Chemical LLC Degarelix acetate P1094-5MG
SDP
Supplier:  Selleck Chemical LLC P10945MG
Encompass_Preferred
Degarelix acetate (ASP-3550 acetate) is a potent and long-acting luteinizing hormone-releasing hormone (LHRH) antagonist
Catalog No. 50-000-43604