Learn More
Essendant Mr. Clean Professional Degreasing Floor Cleaner, 1 gal Bottle, 3/Carton
Supplier:  Essendant PGC19892
Encompass_Preferred
Mr Clean Professional Degreasing Floor Cleaner is a powerful cleaner on greasy floors in commercial kitchens auto-shops concrete and more Has five times the oil and grease cutting power with five concentrated grease cutting actives Mr Clean Degreasing Floor Cleaner is a concentrate that dilutes to make up to 128 gallons of floor cleaning solution to be used with a mop and bucket It can also be used for other washable non-finished surfaces like walls restroom floors tables and on food-contract surfaces Must use appropriate sanitizer after washing Application- Degreaser/Cleaner Floor Maintenance Applicable Material- Floors Food Contact Surfaces Quarry Tile Steel Chemical Compound- Water (7732-18-5) Sodium C10-16 Alkylbenzenesulfonate (68081-81-2) MEA C10-16 Alkylbenzenesulfonate (68910-32-7) Sodium Cumenesulfonate (28348-53-0) Ethanolamine (141-43-5) Trisodium Dicarboxymethyl Alaninate Fragrances Colorants Dirt Types- Grease
Catalog No. 50-000-29238