Learn More
Essendant Dawn Professional Multi-Surface Heavy Duty Degreaser, Fresh Scent, 1 gal Bottle, 2/Carton
Supplier:  Essendant PGC07307
Encompass_Preferred
More Power Less Scrubbing Introducing Dawn Professional Multi-Surface Heavy Duty Degreaser with stronger grease cleaning formula This degreaser cuts through tough grease fast and is safe for food contact surfaces when followed by rinsing with water Great for tough buildup on surfaces like range hoods ovens walls floors drive-thru pads and fuel islands Application- Degreaser/Cleaner Applicable Material- Corral Area Drive-Thru Pads Dumpsters Fryers Hard Surfaces Ovens Parking Lots Range Hoods Sidewalks Walls Chemical Compound- Water Sodium Lauryl Sulfate Lauramine Oxide Sodium Laureth Sulfate Denatured Alcohol Phenoxyethanol Sodium Chloride PPG-26 PEG-8 Propylheptyl Ether Methylisothiazolinone Fragrance Yellow 5 Blue 1 Dirt Types- Greasy Soils Heavy Cooking Grease/Oil Polyerized Animal Grease
Catalog No. 50-000-30067