Learn More
Medchemexpress LLC Dehp-d38 | 352431-42-6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB1945S15MG
Encompass_Preferred
DEHP-d38 is the deuterium labeled DEHP, which is an endogenous metabolite. This compound is useful as a tracer and as an internal standard for quantitative analysis.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-11086