Learn More
Medchemexpress LLC DEHP-d4 (Bis(2-ethylhexyl) phthalate-d4) | 93951-87-2 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB1945S50MG
Encompass_Preferred
DEHP-d4 is the deuterium labeled DEHP. DEHP (Bis(2-ethylhexyl) phthalate) is an endogenous metabolite.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-11205