Learn More
Medchemexpress LLC HY-B1211 500mg Medchemexpress, Dehydroacetic acid CAS:520-45-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1211500MG
Encompass_Preferred
Medchemexpress, HY-B1211 500mg Dehydroacetic acid CAS:520-45-6 Dehydroacetic acid (Biocide 470F), a pyrone derivative acts as an antibacterial and antifungal agent. Dehydroacetic acid possess phytotoxic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2693