Learn More
Medchemexpress LLC Dehydroascorbic acid (Standard) | 490-83-5 | 99.8% | 174.11 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY110281R10MG
Encompass_Preferred
Dehydroascorbic acid (Standard) is the analytical standard of Dehydroascorbic acid. This product is intended for research and analytical applications. Dehydroascorbic acid, a blood-brain barrier transportable form of vitamin C, mediates potent cerebroprotection in experimental stroke.
  • Qualitative research experiments in HPLC, GC and MS.
  • Quantitative research experiments in HPLC, GC and MS.
  • Methodological research experiments in HPLC, GC and MS.
Catalog No. 50-003-43976