Learn More
Medchemexpress LLC Dehydrocholic acid sodium | 145-41-5 | 99.38% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0998500MG
Encompass_Preferred
Dehydrocholic acid sodium is an orally active hydrocholeretic agent that modulates autophagy and reduces serum amylase and lipase levels. It promotes choleretic function, protects the liver, reduces pancreatic damage, and regulates cholesterol metabolism. This product can be used in the study of acute biliary pancreatitis and obstructive jaundice.
  • Modulates autophagy
  • Reduces serum amylase and lipase levels
  • Promotes choleretic function
  • Protects the liver
  • Reduces pancreatic damage
  • Regulates cholesterol metabolism
  • Used in studies of acute biliary pancreatitis
  • Used in studies of obstructive jaundice
Catalog No. 50-004-07457