Learn More
TARGETMOL CHEMICALS INC Dehydrocorydaline chloride 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T109905MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Dehydrocorydaline chloride (13-Methylpalmatine chloride) is an alkaloid exhibiting anti-inflammatory and anti-cancer activities and can enhance the activation of p38 MAPK. Purity 99.88%
Catalog No. 50-287-9494