Learn More
Medchemexpress LLC Dehydrocorydaline nitrate | 13005-09-9 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN42385MG
Encompass_Preferred
Dehydrocorydaline nitrate (13-Methylpalmatine nitrate) is an alkaloid that regulates protein expression of Bax, Bcl-2; activates caspase-7, caspase-8, and inactivates PARP. It elevates p38 MAPK activation and demonstrates anti-inflammatory and anti-cancer activities. It also shows strong anti-malarial effects with an IC50 of 38 nM against P. falciparum 3D7 strain, and low cytotoxicity (cell viability > 90%).
  • Regulates protein expression of Bax, Bcl-2.
  • Activates caspase-7, caspase-8.
  • Inactivates PARP.
  • Elevates p38 MAPK activation.
  • Exhibits anti-inflammatory activities.
  • Exhibits anti-cancer activities.
  • Shows strong anti-malarial effects (IC50 = 38 nM) against P. falciparum 3D7 strain.
  • Low cytotoxicity (cell viability > 90%).
Catalog No. 50-004-19437