Learn More
Medchemexpress LLC Dehydrothio-p-toluidine | 92-36-4 | MFCD00005780 | 99.9% | 240.32 | C14H12N2S | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW040269
Encompass_Preferred
Dehydrothio-p-toluidine (DHPT) is a chemical intermediate used primarily in the production of dyestuffs and related chemical syntheses. The material is supplied as a high-purity solid in laboratory pack sizes and exhibits inhalation toxicity; handle with appropriate engineering controls and personal protective equipment.
  • Chemical intermediate for dye manufacture and synthesis.
  • High purity, suitable for research and development uses.
  • Available in small laboratory pack sizes for convenience.
  • Solid form with specified storage and handling precautions.
  • Requires respiratory protection and controlled handling due to inhalation toxicity.
Catalog No. 50-002-09315