Learn More
Medchemexpress LLC Dehydrotumulosic acid | 6754-16-1 | 1 MG

Supplier: Medchemexpress LLC HYN24991MG
Dehydrotumulosic acid is an active component found in *Poria cocos*, a well-known Chinese medicinal herb. *Poria cocos* has been traditionally used for its abilities to induce diuresis, expel dampness, strengthen the spleen, and calm the mind. Modern pharmacological research also indicates its use in modulating the body's immune system. Dehydrotumulosic acid is provided as a white to off-white solid with a molecular weight of 484.71 and a chemical formula of C31H48O4. It is intended for research use only and is not sold to patients.
- Derived from *Poria cocos*, a traditional Chinese medicinal herb of fungal origin.
- Traditional uses of *Poria cocos*: Induces diuresis, excretes dampness, invigorates the spleen, and tranquilizes the mind.
- Modern pharmacological use: Modulates the immune system.
- Purity: 99.06%
- Form: Solid, white to off-white in color.
- Solubility: Soluble in DMSO at 33.33 mg/mL.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.