Learn More
ATCC Deinococcus radiodurans
Supplier:  ATCC 13939
Encompass
Deinococcus radiodurans strain R1 (smooth) was isolated from irradiated ground pork and beef. This bacterial strain is radiation-resistant and is known to produce restriction endonuclease MraI.
Catalog No. 50-238-2588