Learn More
Peak Scientific Instruments Ltd DEIONIZER CARTRIDGE SPARE PART
SDP
Supplier:  Peak Scientific Instruments Ltd 057062
Encompass_Preferred
  • Designed for hydrogen gas generation.
  • Compatible with Peak Scientific Precision Hydrogen Series (all color screen modules).
  • Provides deionized water to meet ASTM Type II purity standards (<1.0 uS / > 1MΩ).
  • Contributes to high purity hydrogen gas production (up to 99.9995%).
  • Essential for routine maintenance, along with silica gel.
  • Certified CSA approved.
  • Included in precision hydrogen service kits.
Catalog No. NC1184510