Learn More
Biocare Medical, LLC Eosin-Phloxine - 1 Gallon
SDP
Supplier:  Biocare Medical, LLC HTEPGL
Encompass_Preferred
This is a specially formulated alcoholic Eosin Y/Phloxine B counterstain. This Eosin-Phloxine has been modified with the addition of stabilizers to prolong shelf life. Staining intensity is more subtle than other Eosin Y/Phloxine B formulations, however, the pink shades are more vivid and alcoholic hyaline is also stained.
Catalog No. NC1441271