Learn More
Cayman Chemical DelafloxacIn 5mg
SDP
Supplier:  Cayman Chemical 203145 MG
Encompass_Preferred
A fluoroquinolone antibiotic that demonstrates extremely high potency against a wide range of bacteria in vitro; has an anionic structure, which enhances its potency in acidic environments and in skin and soft tissue infections
Catalog No. 50-298-2811