Learn More
Medchemexpress LLC HY-10571A 100mg Medchemexpress, Delavirdine (mesylate) CAS:147221-93-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10571A100MG
Encompass_Preferred
Medchemexpress, HY-10571A 100mg Delavirdine (mesylate) CAS:147221-93-0 Delavirdine (U 90152; BHAP-U 90152) mesylate is a potent, highly specific and orally active non-nucleoside reverse transcriptase inhibitor (NNRTI). Delavirdine mesylate selectively inhibits HIV-1 reverse transcriptase (RT) (IC50=0.26 μM) over DNA polymerase α (IC50=440 μM) and polymerase δ (IC50>550 μM). Delavirdine mesylate is an inhibitor of HIV-1 replication and can can be used for the study of AIDs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8770
 Restricted Item