Learn More
ATLANTIC BIOPHARM SOLUTIONS LLC DELIVERY HOSE W/CAST TRICLAMP
Supplier:  ATLANTIC BIOPHARM SOLUTIONS LLC DG003852
Encompass_Preferred
Delivery hose with cast TriClamp AIRDUC PUR 350 FOOD-AS Ø-Inner 1.25"/ 32mm L=2000mm
Catalog No. NC2871878