Learn More
Bellco Glass, Inc. Delong 3-Baffles Nephelo,300mL 19x130 Side Arm
SDP
Supplier:  Bellco Glass, Inc. 257819133
Encompass_Preferred
Ideal for growing, storing and mixing or aerobic and anaerobic cultures. The 3 bottom baffles increase culture oxygenation. Includes 25mm Bellco stainless steel closure (2005 series). MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 300mlIncludes: Bellco Stainless Steel Closure (2005 Series)Sidearm Dimensions: 19x130mmCase Quantity: 4
Catalog No. 50-194-5582