Learn More
Bellco Glass, Inc. DeLong 3 Baffles Fernbach,1800mL Bottom Baffles
SDP
Supplier:  Bellco Glass, Inc. 255401800
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Designed for culturing organisms that require a large surface area to volume ratio. Features a 38mm DeLong style neck and can be used with a 38mm stainless steel closure (sold separately). MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 1800mlOD x OAH: 200 x175mmCase Quantity: 3
Catalog No. 50-194-5562