Learn More
Bellco Glass, Inc. Delong Fernbach Flask, 1800mL No Baffles
SDP
Supplier:  Bellco Glass, Inc. 255001800
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Ideal for culturing organisms requiring a large surface to volume. DeLong style neck and uses 38mm stainless steel closure (sold separately). MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 1800mlOD x OAH: 200 x 175mmCase Quantity: 3
Catalog No. 50-194-5549