Learn More
Bellco Glass, Inc. DeLong Flask Complete, 2L
SDP
Supplier:  Bellco Glass, Inc. 251002000
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for growing, storing and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes. MADE IN THE USA Size: 2l Closure: 38 Dimensions: 158x295mm
Catalog No. 50-194-5458