Learn More
Bellco Glass, Inc. DeLong Flask Only,6L
SDP
Supplier:  Bellco Glass, Inc. 251106000
Encompass_Preferred
Manufactured for type 1 class a Borosilicate glass. Ideal for growing, storing, and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes.. MADE IN THE USA Size: 6l Closure: 38 Dimensions: 235x410mm Stainless steel closure or Kaput closure sold separately
Catalog No. 50-194-5471