Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Delphinidin 3-O-beta-D-galactoside chloride | 28500-00-7 | MFCD05864894 | 98.7% | 500.84 g/mol | C21H21ClO12 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN660610MG

Encompass_Preferred

Delphinidin 3-O-beta-D-galactoside chloride is an anthocyanin analytical standard used for antioxidant and antimicrobial research and for analytical method development. It is supplied as a purified chloride salt suitable for HPLC reference standards, identity confirmation, quantitative assays, and bioactivity testing.

  • High purity (98.7%).
  • Chemical formula C21H21ClO12.
  • Molecular weight 500.84 g/mol.
  • CAS number 28500-00-7.
  • Suitable as an HPLC analytical standard and reference material.
  • Available in 10 mg packaging for analytical use.

Catalog No. 50-002-89524


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.