Learn More
Medchemexpress LLC Delphinidin 3-O-glucoside chloride | 6906-38-3 | MFCD01762899 | 99.9% | 500.84 | C21H21ClO12 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10805210MM 1ML
Encompass_Preferred
Delphinidin 3-glucoside chloride is an anthocyanin reference standard supplied as a ready-to-use 10 mM solution in DMSO (1 mL). It is used in biochemical and cell biology research to investigate antioxidant activity, cell signaling, apoptosis, and endothelial function. The compound has molecular formula C21H21ClO12, molecular weight 500.84, and high purity suitable for analytical work.
  • Provided as a 10 mM solution in DMSO, 1 mL.
  • High purity (99.89%) suitable for analytical and biological assays.
  • Well-characterized molecular formula C21H21ClO12 and molecular weight 500.84.
  • Ready for immediate use in cell signaling and oxidative stress studies.
  • Stable when stored according to supplier recommendations.
Catalog No. 50-002-43061