Learn More
STEMCELL Technologies Delphinidin (Chloride), Size: 1 mg
SDP
Supplier:  STEMCELL Technologies 74232
Encompass
Delphinidin is an anthocyanidin found in pigmented fruits and vegetables with anti-inflammatory, anti-angiogenic, and anti-oxidant properties (Watson & Schönlau). It has been reported to be a potent inhibitor of epidermal growth factor receptor (EGFR; Meiers et al.) and vascular endothelial growth factor receptor (VEGFR) (Lamy et al.). This product is supplied as the chloride salt of the molecule.CANCER RESEARCH· Induces G2-M phase arrest in human prostate cancer PC3 cells, leading to apoptosis (Hafeez et al.).
Catalog No. 50-197-6202