Learn More
Cayman Chemical DelphInIdIn3OrutInosIdec 5mg
SDP
Supplier:  Cayman Chemical 335525 MG
Encompass_Preferred
A glycosylated anthocyanin with antioxidant activity; inhibits TBHP-induced loss of viability in isolated rat aortic SMCs, but not MH1C1 cells, at 100 µM; prevents TBHP-induced DNA damage, as well as inhibits TBHP-induced increases in MDA levels, in SMCs; displays a hydrogen peroxide-induced and pH-dependent chemiluminescence peak at pH 7
Catalog No. 50-294-6562