Learn More
Cayman Chemical DelphInIdInchlorIde 10mg
SDP
Supplier:  Cayman Chemical 1101210 MG
Encompass_Preferred
A natural plant pigment which induces the release of nitric oxide by vascular endothelium, causing vasorelaxation; inhibits signaling through EGFRs, suppressing the expression of ERα and inducing both apoptosis and autophagy at a dose of 1-40 μM; inhibits the HAT activities of p300/CBP (IC50 = ~30 μM)
Catalog No. 50-295-1642