Learn More
Cytiva T-series Centramate cassette DELTA regenerated cellulose membrane 10 kDa MWCO 93 cm
Supplier:  Cytiva DC010T01
Encompass
T-series Centramate cassette DELTA regenerated cellulose membrane 10 kDa MWCO 93 cm
Catalog No. 50-000-52780