Learn More
Cytiva T-series Centrasette cassette DELTA regenerated cellulose membrane 30 kDa MWCO 2 5 m
Supplier:  Cytiva DC030T26
Encompass
T-series Centrasette cassette DELTA regenerated cellulose membrane 30 kDa MWCO 2 5 m
Catalog No. 50-000-52836
Only null left
Add to Cart
Request a Quote