Learn More
Medchemexpress LLC Delta-nonalactone | 3301-94-8 | MFCD00036500 | 99.3% | 156.22 g/mol | C9H16O2 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW017284
Encompass_Preferred
Delta-nonalactone (δ-nonalactone) is a cyclic lactone (C9H16O2, CAS 3301-94-8) used as a flavor and fragrance ingredient and in laboratory research. It is a colorless to pale yellow liquid with a coconut-like odor and is supplied at high purity for analytical and experimental use.
  • High purity appropriate for research and analytical use.
  • Coconut-like aroma for flavor and fragrance formulation.
  • Reported insect-repellent activity against tsetse flies.
  • Colorless to pale yellow liquid at ambient conditions.
  • Density approximately 0.994-0.999 g/mL.
  • Used in biochemical assays and metabolic research.
Catalog No. 50-003-02322