Learn More
Waters Corp Waters Column, Delta-Pak C4, 5 μm, 100 Å, 3.9 x 150 mm
Supplier: Waters Corp WAT011796
Delta-Pak packings, based on highly stable, bonded, endcapped 5μm spherical silica, are designed for legacy methods. This reversed-phase HPLC column features a C4 bonded phase and a 100Å pore size, facilitating consistent scale-up and easy purity checks. It operates from pH 2 to 8, with a maximum pressure of 6000 psi (415 Bar), and supports molecular weights from 1000 to 15000.
- Designed for legacy methods
- Features C4 bonded phase and 100Å pore size
- Consistent scale-up and easy purity checks
- pH range of 2 to 8; max pressure 6000 psi (415 Bar)
- Suitable for molecular weights from 1000 to 15000
- Low silanol activity, 7% carbon load
- Dimensions: 3.9 mm ID × 150 mm L
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.