Learn More
CPC Scientific H-Trp-Ala-Gly-Gly-Asp-Ala-Ser-Gly-Glu-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific DSIP001A
Encompass
A neuropeptide that when infused into the mesodiencephalic ventricle of recipient rabbits induces spindle and delta EEG activity and reduced motor activitiesONE-LETTER SEQUENCE: WAGGDASGEMOLECULAR FORMULA: C35H48N10O15MOLECULAR WEIGHT:848.82STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [62568-57-4], RESEARCH AREA: NeuropeptidesREFERENCES: G.A.Schoenenberger and M.Monnier, Proc. Natl. Acad. Sci. USA, 74, 1282 (1977)
Catalog No. 50-210-9199