Learn More
Cayman Chemical δ-Tocotrienol, 25612-59-3, 1 mg
SDP
Supplier:  Cayman Chemical 100085131
Encompass_Preferred
At 0.1-10 µM, .delta.-tocotrienol significantly suppresses H2O2-induced cell death of rat striatal neurons and provides protection against S-nitrocysteine-induced cell death and SIN-1 cytotoxicity in a concentration dependant manner.{14290} At concentrations of 10 µM, .delta.-tocotrienol displays more inhibitory action on adhesiveness of HAECs to U937 monocytic cells, via inhibition of VCAM-1 expression, than α-tocopherol and other tocotrienols at the same concentration.{14291}
Catalog No. NC0104799