Learn More
Medchemexpress LLC HY-114202 10mg Medchemexpress, delta-Valerobetaine CAS:6778-33-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11420210MG
Encompass_Preferred
Medchemexpress, HY-114202 10mg delta-Valerobetaine CAS:6778-33-2 delta-Valerobetaine is a precursor of trimethylamine N-oxide (TMAO). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9305