Learn More
Medchemexpress LLC Deltamethrin-d5 (mixture of diastereomers) | 1217633-23-2 | MFCD09840317 | >=99.0% | 510.23 g/mol | C22H14D5Br2NO3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW6537691MG
Encompass_Preferred
Deltamethrin-d5 (mixture of diastereomers) is the deuterium-labeled analogue of deltamethrin supplied as an isotopically labeled reference material for analytical and metabolic studies.
  • Isotopically labeled with five deuterium atoms (d5).
  • Supplied as a mixture of diastereomers.
  • Molecular formula: C22H14D5Br2NO3; molecular weight: 510.23 g/mol.
  • Available in small mass quantities suitable for analytical standards (1 mg, 5 mg, 10 mg).
  • Commonly used as an internal standard for GC- or LC-MS quantification and metabolic tracing.
Catalog No. 50-002-83915