Learn More
Medchemexpress LLC Deltasonamide 1 | 2088485-33-8 | 99.2% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12264110MG
Encompass_Preferred
Deltasonamide 1 is a small-molecule inhibitor of PDE6δ-KRas with high affinity (KD ≈ 203 pM) and is supplied for research applications, including tumor research. It is provided as a solid and in DMSO solution formulations with recommended storage guidance.
Catalog No. 50-002-31614