Learn More
Medchemexpress LLC Deltasonamide 1 (TFA) | 2235358-73-1 | 99.7% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY122641D1MG
Encompass_Preferred
Deltasonamide 1 TFA is a small-molecule research compound that inhibits the PDE6δ-KRas interaction with a reported KD of 203 pM. It is supplied as the trifluoroacetate salt for laboratory research and is characterized by a molecular weight of 761.27 g/mol and CAS 2235358-73-1.
  • High-affinity PDE6δ-KRas inhibitor (KD ≈ 203 pM).
  • Supplied as a trifluoroacetate salt for improved handling.
  • Purity 99.7% suitable for research applications.
  • Available in milligram quantities for screening and biochemical assays.
  • Useful for tumor and Ras-pathway research.
Catalog No. 50-002-31613