Learn More
Medchemexpress LLC Deltasonamide 2 hydrochloride | 2448341-55-5 | 99.8% | 1 ML

Supplier: Medchemexpress LLC HY122641C10MM 1ML
Deltasonamide 2 hydrochloride is the hydrochloride salt of a small-molecule inhibitor that binds phosphodiesterase δ (PDEδ) with high affinity (Kd ≈ 385 pM). It is supplied as a solid or as a ready-to-use 10 mM solution in DMSO for research use; handle according to safety documentation.
- Competitive, high-affinity PDEδ inhibitor (Kd ≈ 385 pM).
- Hydrochloride salt to improve solubility for biological assays.
- Available as solid and as a 10 mM solution in DMSO (1 mL).
- High purity, typically 99.78%.
- Molecular weight 683.71 g/mol; formula C30H40Cl2N6O4S2.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.