Learn More
CPC Scientific H-Tyr-DMet-Phe-His-Leu-Met-Asp-NH2 (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific OPIO010B
Encompass
Deltorphin, YmFHLMD-amide, a peptide first isolated from skin extracts of the South American frog Phyllomedusa sauvagei, corresponds to the predicted prodermorphin heptapepeptide. Deltorphin acts as a highly potent and fully selective agonist for the delta-opioid receptor.ONE-LETTER SEQUENCE: Y-DMet-FHLMD-NH2MOLECULAR FORMULA: C44H62N10O10S2MOLECULAR WEIGHT:955.17STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [119975-64-3], RESEARCH AREA: PainREFERENCES: M.Amiche et al., Mol. Pharmacol., 35, 774 (1989); G.Kreil et al., Eur. J. Pharmacol., 162, 123 (1989); L.H.Lazarus et al., J. Biol. Chem., 264, 3047 (1989); A.Mor et al., FEBS Lett., 255, 269 (1989); S.Sagan et al., J. Biol. Chem., 264, 17100 (1989); P.Melchiorri et al., Eur. J. Pharmacol., 195, 201 (1991); S.Salvadori et al., J. Med. Chem., 34, 1656 (1991); L.H.Lazarus et al., J. Med. Chem., 34, 1350 (1991)
Catalog No. 50-210-9983