Learn More
Selleck Chemical LLC Demecarium Bromide S9463-100MG
SDP
Supplier:  Selleck Chemical LLC S9463100MG
Encompass_Preferred
Demecarium Bromide an anticholinesterase is an inhibitor of monoamine oxidase (MAO) It induces long-term miosis and decreases intraocular pressure (IOP) in normotensive eyes of Beagles and of eyes of Beagles with inherited glaucoma
Catalog No. 50-000-23988