Learn More
Medchemexpress LLC Demethoxycurcumin-d4 | 22608-11-3 | MFCD03427310 | 98.9% | 342.38 g/mol | C20H14D4O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0006S110MG
Encompass_Preferred
Demethoxycurcumin-d4 is the deuterium-labeled analog of demethoxycurcumin used as an isotopically labeled reference standard in biochemical and pharmacological research. It is suitable for use as an internal standard in mass spectrometry, tracer studies, and quantitative assays where a stable isotope label improves accuracy and reduces matrix effects.
  • Isotopically labeled internal standard for quantitation.
  • High reported purity suitable for analytical applications.
  • Soluble in DMSO at high concentration with ultrasonic aid.
  • Stable when stored as powder at recommended temperatures.
  • Useful in studies of inflammation, apoptosis, and metabolic profiling.
Catalog No. 50-002-97710