Learn More
Medchemexpress LLC Demethoxycurcumin-d4 (Curcumin II-d4) | 22608-11-3 | 98.9% | 342.38 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN0006S11MG
Encompass_Preferred
Demethoxycurcumin-d4 is the deuterium labeled Demethoxycurcumin. Demethoxycurcumin (Curcumin II) is a major active curcuminoid with anti-inflammatory properties. It can be used as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Application as tracer
  • Internal standard for quantitative analysis
  • Deuterium substitution affects pharmacokinetic and metabolic profiles
Catalog No. 50-004-28744